Calcium bis(dihydrogenorthophosphate) monohydrate ≥99.0%, BAKER ANALYZED®, J.T.Baker®

Synonyms: Calcium phosphate, monobasic, monohydrate

Management of Change (MOC) category = N

BAKR1426-05 BAKR1426-01 1426-05 BAKR1426-01
JT1426-1CS 3212.96 USD
JT1426-1 CAJT1426-5 JT1426-5 CAJT1426-1
Calcium bis(dihydrogenorthophosphate) monohydrate ≥99.0%, BAKER ANALYZED®, J.T.Baker®
Calcium bis(dihydrogenorthophosphate) monohydrate

Crystals. Lot analysis on label.

Formula: H₄CaO₈P₂·1H₂O
MW: 252.07 g/mol
MDL Number: MFCD00149602
CAS Number: 10031-30-8
EINECS: 231-837-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (Ca(H₂PO₄)₂·H₂O) 99 - 105 %
Insoluble in HCl ≤0.01 %
Chloride (Cl) ≤0.002 %
Sulfate (SO₄) ≤0.01 %
Ammonium (NH₄) ≤0.01 %
Heavy Metals (as Pb) ≤0.002 %
Iron (Fe) ≤0.002 %
Trace Impurities - Arsenic (As) ≤1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR