Calcium fluoride ≥95.0% (by EDTA titration), powder, BAKER ANALYZED®, J.T.Baker®


Management of Change (MOC) category = N

BAKR1354-01 BAKR1354-07 BAKR1354-01
JT1354-1CS 2908.41 USD
JT1354-1 CAJT1354-1 JT1354-07
Calcium fluoride ≥95.0% (by EDTA titration), powder, BAKER ANALYZED®, J.T.Baker®
Calcium fluoride

Powder.

Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: 2500 °C (1013 hPa)
Melting Pt: 1418 °C
Density: 3,18 g/cm³ (20 °C)
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CaF₂) (by EDTA titrn) ≥95.0 %
Chloride (Cl) ≤0.1 %
Sulfate (SO₄) ≤0.05 %
Heavy Metals (as Pb) ≤0.005 %
Iron (Fe) ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR