Calcium carbonate ≥98%, light powder FCC, USP, J.T.Baker®

Synonyms: Marble

Management of Change (MOC) category = R

BAKR1301-05 1301-05 BAKR1301-01 1301-01 BAKR1301-QM
JT1301-5EA 917.28 USD
JT1301-5 CAJT1301-1 JT1301-QM JT1301-1 CAJT1301-5
Calcium carbonate ≥98%, light powder FCC, USP, J.T.Baker®
Calcium carbonate

.

Formula: CaCO₃
MW: 100.09 g/mol
Melting Pt: 825 °C
Density: 2.93 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Order Now

Specification Test Results

Meets F.C.C. Requirements
Meets U.S.P Requirements
GMP Manufactured Product
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (CaCO₃) (dried basis) 98.0 - 100.5 %
USP - Identification Passes Test
USP - Acid-Insoluble Substances ≤0.2 %
USP - Fluoride (F) ≤0.005 %
USP - Arsenic (As) ≤3 ppm
USP - Barium (Ba) Passes Test
USP - Iron (Fe) ≤0.1 %
USP - Lead (Pb) ≤3 mg/kg
USP - Magnesium and Alkali Salts ≤1.0 %
USP - Mercury (Hg) ≤0.5 ppm
FCC - Assay (CaCO₃) (dried basis) 98.0 - 100.5 %
FCC - Identification Passes Test
FCC - Loss on Drying ≤2.0 %
FCC - Acid-Insoluble Matter ≤0.2 %
FCC - Fluoride (F) ≤0.005 %
FCC - Arsenic (As) ≤3 ppm
FCC - Lead (Pb) ≤3 mg/kg
FCC - Magnesium and Alkali Salts ≤1.0 %
Bulk Density (lb/cu ft) 32 - 36
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR