Cadmium(II) chloride hemipentahydrate 79.5-81.0% (by argentometric titration), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

BAKR1208-01
JT1208-1CS 5208.83 USD
JT1208-1
Cadmium(II) chloride hemipentahydrate 79.5-81.0% (by argentometric titration), BAKER ANALYZED® ACS, J.T.Baker®
Cadmium (II) chloride hemipentahydrate

Crystals.

Formula: CdCl₂·2.5H₂O
MW: 228.35 g/mol
Melting Pt: 568 °C
Density: 2.84 g/cm³ (20 °C)
MDL Number: MFCD00799071
CAS Number: 7790-78-5
EINECS: 233-296-7
UN: 2570
ADR: 6.1,III
Merck Index: 13,01617

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CdCl₂) (by Ag titrn) 79.5 - 81.0 %
Insoluble Matter ≤0.005 %
Nitrate and Nitrite (as NO₃) ≤0.003 %
Sulfate (SO₄) ≤0.005 %
Ammonium (NH₄) ≤0.002 %
Calcium (Ca) ≤0.005 %
Lead (Pb) ≤0.005 %
Potassium (K) ≤0.02 %
Sodium (Na) ≤0.05 %
Zinc (Zn) ≤0.05 %
Trace Impurities - Copper (Cu) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR