Cyclopentane, OmniSolv®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Pentamethylene
CX2414-1
CX2414-1
EM-CX2414-1EA
675
USD
EM-CX2414-1
CACX2414-1
Cyclopentane, OmniSolv®, Supelco®
Cyclopentane
For spectrophotometry, liquid chromatography, and residue analysis. UV cutoff 198nm. Filtered through a 0.2µm element. Lot analysis on label.
Formula:
C₅H₁₀ MW: 70.13 g/mol Boiling Pt: 49 °C (1013 hPa) Melting Pt: –93.3 °C Density: 0.746 g/cm³ (20 °C) Flash Pt: –41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001356 CAS Number: 287-92-3 EINECS: 206-016-6 UN: 1146 ADR: 3,II Merck Index: 13,02769 |
Specification Test Results
Assay (2,2-Dimethylbutane) | 15.0% max |
Assay (Cyclopentane) | 85.0% min |
Assay (n-Pentane) | 3.0% max |
Assay (Total) | 99.8% min |
Capillary ECD responsive substances (as PCNB) | 5ng/L max |
Color (APHA) | 10 max |
Filtered through 0.2 µm filter | To pass test |
Fluorescence (as quinine base) | 500ppt max |
Form | Clear liquid |
Infrared spectrum | Conforms to standard |
Residue after evaporation | 1ppm max |
UV Abs. at 198 nm | 1.00AU max |
UV Abs. at 210 nm | 0.50AU max |
UV Abs. at 220 nm | 0.10AU max |
UV Abs. at 230 nm | 0.01AU max |
UV Cut-off | 198nm max |
Water (H2O) | 0.02% max |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...