Xylenol orange tetrasodium salt, Laboratory Reagent, J.T.Baker®

Synonyms: Xylenol orange water soluble

Management of Change (MOC) category = N

BAKRX584-02 BAKRX584-01 X584-01 X584-02
JTX584-1EA 221.35 USD
JTX584-1 JTX584-2 CAJTX584-2 CAJTX584-1
Xylenol orange tetrasodium salt, Laboratory Reagent, J.T.Baker®
Xylenol orange tetrasodium salt
Formula: C₃₁H₂₈N₂Na₄O₁₃S
MW: 760.59 g/mol
Melting Pt: ∼210 °C
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS Number: 3618-43-7
EINECS: 222-805-8

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Clarity of Solution Passes Test
Suitability for Zinc Titration Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR