Butyl-2-bromopropionate ≥95.0%
Supplier: TCI America
Synonyms:
2-Bromopropionic Acid Butyl Ester
B1031-25ML
TCB1031-025MLEA
31.45
USD
TCB1031-025ML
Butyl-2-bromopropionate ≥95.0%
Butyl-2-bromopropionate
CAS Number: 41145-84-0
MDL Number: MFCD00059469
Molecular Formula: C7H13BrO2
Molecular Weight: 209.08
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 196
Specific Gravity (20/20): 1.28
Other Product Names / Synonyms: 2-Bromopropionic Acid Butyl Ester
Formula:
C₇H₁₃BrO₂ MW: 209.08 g/mol Boiling Pt: 196 °C Density: 1.28 (20 °C) |
MDL Number:
MFCD00059469 CAS Number: 41145-84-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...