2-Butoxyethanol ≥99% (by GC), J.T.Baker®
|
Danger
|
Synonyms:
Ethylene glycol monobutyl ether
, Ethylene glycol butyl ether
, Butyl cellosolve
Management of Change (MOC) category = N
BAKRD648-09
BAKRD648-07
BAKRD648-07
BAKRD648-09
JTD648-9CS
768.7
USD
JTD648-9
JTD648-7
CAJTD648-9
CAJTD648-7
2-Butoxyethanol ≥99% (by GC), J.T.Baker®
2-Butoxyethanol
Formula:
CH₃(CH₂)₃OCH₂CH₂OH MW: 118.18 g/mol Boiling Pt: 171 °C (1013 hPa) Melting Pt: –70 °C Density: 0.902 g/cm³ (20 °C) Flash Pt: 61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002884 CAS Number: 111-76-2 EINECS: 203-905-0 UN: 2810 ADR: 6.1,III Merck Index: 13,01558 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (CH₃(CH₂)₃OCH₂CH₂OH) (by GC) | ≥99 % |
Color (APHA) | ≤10 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...