Bromophenol blue, BAKER ANALYZED® ACS, J.T.Baker®

Synonyms: Bromphenol Blue Sultone FormTetrabromophenol-sulfonephthalein3,3',5,5'-TetrabromophenolsulfonphthaleinTetrabromophenol-sulphonephthaleinBPB3,3',5,5'-Tetrabromophenolsulphonphthalein

Management of Change (MOC) category = N

BAKRD293-03 D293-03
JTD293-3EA 217.42 USD
CAJTD293-3 JTD293-3
Bromophenol blue, BAKER ANALYZED® ACS, J.T.Baker®
Bromophenol blue

Lot analysis on label.

Formula: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
Melting Pt: 273 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2
Merck Index: 13,01429

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Clarity of Solution Passes Test
Visual Transition Interval - pH…4.6 (Blue) Passes Test
Visual Transition Interval - pH…3.0 (Yellow) Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR