Bis(2-ethylhexyl)phosphate ≥95%, J.T.Baker®

Danger

Synonyms: Di-(2-ethylhexyl)phosphoric acidBis(2-ethylhexyl) hydrogen phosphate

BAKRC533-07
CAJTC533-7
Bis(2-ethylhexyl)phosphate ≥95%, J.T.Baker®
Bis(2-ethylhexyl) phosphate
Formula: C₁₆H₃₅O₄P
MW: 322.43 g/mol
MDL Number: MFCD00009492
CAS Number: 298-07-7
EINECS: 206-056-4
UN: 1902
ADR: 8,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₁₆H₃₅O₄P) ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR