Boric acid, ULTREX®, Ultrapure, J.T.Baker®

Danger

Synonyms: Boric acidBoron hydroxideBoron trihydroxideBoracic acidOrthoboric acid

Management of Change (MOC) category = N

BAKR5168-05 BAKR5168-05
JT5168-5EA 491.22 USD
CAJT5168-5 JT5168-5
Boric acid, ULTREX®, Ultrapure, J.T.Baker®
ortho-Boric acid

For fiber optics manufacture.

Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://us.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (H₃BO₃) (by acid-base titration), For Information Only
Particulate Matter, For Information Only
Nonvolatile with Methanol, For Information Only
Non-Metallic Impurities - Arsenic (As), For Information Only
Non-Metallic Impurities - Halide (as Cl), For Information Only
Non-Metallic Impurities - Phosphate (PO₄), For Information Only
Non-Metallic Impurities - Sulfur Compounds (as SO₄), For Information Only
Metallic Impurities - Aluminum (Al), For Information Only
Metallic Impurities - Barium (Ba), For Information Only
Metallic Impurities - Calcium (Ca), For Information Only
Metallic Impurities - Chromium (Cr), For Information Only
Metallic Impurities - Cobalt (Co), For Information Only
Metallic Impurities - Copper (Cu), For Information Only
Metallic Impurities - Iron (Fe), For Information Only
Metallic Impurities - Manganese (Mn), For Information Only
Metallic Impurities - Nickel (Ni), For Information Only
Metallic Impurities - Tin (Sn), For Information Only
Metallic Impurities - Titanium (Ti), For Information Only
Metallic Impurities - Vanadium (V), For Information Only
Metallic Impurities - Zirconium (Zr), For Information Only
Certificate Provided Reports Actual Lot Analysis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR