Barium chloride dihydrate ≥99.0% (by EDTA titration), BAKER ANALYZED® ACS, J.T.Baker®

Danger


Management of Change (MOC) category = N

BAKR0970-01 BAKR0970-04 BAKR0970-05 BAKR0970-01
JT0970-5CS 5719.51 USD
JT0970-5 JT0970-4 JT0970-1 CAJT0970-1
Barium chloride dihydrate ≥99.0% (by EDTA titration), BAKER ANALYZED® ACS, J.T.Baker®
Barium chloride dihydrate
Formula: BaCl₂·2H₂O
MW: 244.27 g/mol
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 960 °C
Density: 3.86 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 13,00974

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (BaCl₂·2H₂O) (by EDTA titrn) ≥99.0 %
Loss on Drying at 150°C 14.0 - 16.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 5.2 - 7.0
Oxidizing Substances (as NO₃) ≤0.005 %
Calcium (Ca) ≤0.05 %
Potassium (K) ≤0.0025 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.1 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR