Barium acetate 99.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
|
Warning
|
Synonyms:
Barium di(acetate)
, Acetic acid barium salt
Management of Change (MOC) category = N
BAKR0942-01
BAKR0942-05
JT0942-1CS
698.64
USD
JT0942-1
JT0942-5
Barium acetate 99.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Barium acetate
Formula:
(H₃CCOO)₂Ba MW: 255.42 g/mol Melting Pt: ∼450 °C Density: 2.47 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012447 CAS Number: 543-80-6 EINECS: 208-849-0 UN: 1564 ADR: 6.1,III Merck Index: 12,00992 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay ((CH₃COO)₂Ba) | 99.0 - 102.0 % |
Insoluble Matter | ≤0.01 % |
pH of 5% Solution at 25°C | 7.0 - 9.0 |
Chloride (Cl) | ≤0.001 % |
Oxidizing Substances (as NO₃) | ≤0.005 % |
Calcium (Ca) | ≤0.05 % |
Potassium (K) | ≤0.003 % |
Sodium (Na) | ≤0.005 % |
Strontium (Sr) | ≤0.2 % |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...