ICNA02903312
0290331280 0290331205 0290331225
IC90331225EA 185.34 USD
IC90331225 IC90331205 IC90331280
ICNA02903312
Brewer's yeast is often used as a source of B-complex vitamins, chromium, and selenium. While brewer's yeast can be used to bake bread, ferment wine, beer and ethanol production. Also used as an insect diet.
  • Protein >35%
Brewer's yeast is basically S. cerevisiae, is the same species as the common baking yeast, but rich in essential minerals and the vitamins.