Boron trifluoride - methanol complex 10 - 15% (w/v) in methanol, Supelco®

Danger

BX0917-1 BX0917-1
EM-BX0917-1EA 363.03 USD
EM-BX0917-1 CABX0917-1
Boron trifluoride - methanol complex 10 - 15% (w/v) in methanol, Supelco®
Boron trifluoride methanol complex

For preparation of methyl esters of fatty acids.

Formula: CH₄BF₃O
MW: 99.85 g/mol
MDL Number: MFCD00071635
CAS Number: 373-57-9
EINECS: 206-766-4
UN: 3286
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR