Acrylamide polymer, M.W. M.W.=400,000-800,000 10% in water (containing small amounts of formalin as fungicide)

Supplier: TCI America

Synonyms: Polyacrylamidepoly(1-carbamoylethylene)2-Propenamide Homopolymer

A0140-500G
TCA0140-500GEA 207.82 USD
TCA0140-500G
Acrylamide polymer, M.W. M.W.=400,000-800,000 10% in water (containing small amounts of formalin as fungicide)
Acrylamide polymer

CAS Number: 9003-05-8
MDL Number: MFCD00084392
Form: Starch Syrup
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Polymers

Formula: (CH2:CHCONH2)n
MDL Number: MFCD00084392
CAS Number: 9003-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR