Agarose standard, Ultrapure Bioreagent for electrophoresis low electroendosmosis (EEO), J.T.Baker®

Warning


Management of Change (MOC) category = N

BAKRA426-07 A426-07 A426-05 BAKRA426-05
JTA426-5EA 489.62 USD
CAJTA426-7 JTA426-5 JTA426-7 CAJTA426-5
Agarose standard, Ultrapure Bioreagent for electrophoresis low electroendosmosis (EEO), J.T.Baker®
Agarose

Standard, low electroendosmosis (EEO). For routine protein and nucleic acid electrophoresis. Lot analysis on label.

Melting Pt: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://us.vwr.com/stibo/web/std.lang.all/43/75/23914375.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Loss on Drying at 70°C (in vacuo) ≤7 %
Sulfate (SO₄) ≤0.2 %
Electroendosmosis (EEO)(-m^r) ≤0.13
Gel Strength, g/cm² (MCl, 1.0%) ≥1200
Gelling Temperature of 1.5% w/w Solution 34.5 - 37.5 °C
Melting Temperature of 1.5% w/w Solution 86.5 - 89.5 °C
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR