Acetanilide, BAKER ANALYZED®, J.T.Baker®

Warning

Synonyms: N-Phenylacetamide

Management of Change (MOC) category = N

BAKRA068-03
JTA068-3EA 153.57 USD
JTA068-3
Acetanilide, BAKER ANALYZED®, J.T.Baker®
Acetanilide

Lot analysis on label. Reference standard for determination of carbon, hydrogen, nitrogen, and oxygen.

Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 174 °C
MDL Number: MFCD00008674
CAS Number: 103-84-4
EINECS: 203-150-7
Merck Index: 13,00051

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Carbon (C), For Information Only
Hydrogen (H), For Information Only
Nitrogen (N), For Information Only
Oxygen (O), For Information Only
Appearance (white to off-white, grayish or brownish crystalline scales or crystalline powder)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR