L(+)-Arginine hydrochloride 98.5-101.5% USP, Multi-compendial, J.T. Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS

Synonyms: (S)-(+)-Arginine hydrochlorideL-Arginine , hydrochlorideL-(+)-Arginine MonohydrochlorideL-Arginine monohydrochlorideH-L-Arg-OH.HClH-Arg-OH.HCl

2067-06 2067-06 2067-07 2067-09 2067-07 2067-09
JT2067-6EA 379.81 USD InStock
CAJT2067-6 JT2067-6 CAJT2067-7 CAJT2067-9 JT2067-7 JT2067-9
L(+)-Arginine hydrochloride 98.5-101.5% USP, Multi-compendial, J.T. Baker®
L(+)-Arginine hydrochloride

The J.T.Baker® Global Pharma™ line of biopharmaceutical and pharmaceutical amino acids is cGMP-manufactured to meet the toughest regulatory requirements and tested for each specification of the appropriate compendia.

Formula: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
Melting Pt: 218 °C
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064550
CAS Number: 1119-34-2
EINECS: 214-275-1
Merck Index: 13,00785

Order Now

Specification Test Results

Food GMP Manufactured Product
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Preserve in well-closed containers.
Store in well-closed,light-resistant containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (C₆H₁₄N₄O₂HCl) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotationα²ºD (+) 21.4 - 23.6 Degree
USP - Loss on Drying at 105°C ≤ 0.2 %
USP - Residue on Ignition ≤ 0.1 %
USP - Sulfate (SO₄) ≤ 0.03 %
USP - Related Compounds - Individual Impurities ≤ 0.5 %
USP - Related Compounds - Total Impurities ≤ 2.0 %
USP - Chloride (Cl) 16.5 - 17.1 %
EP/BP - Assay (C₆H₁₄N₄O₂HCl) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification E Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotationα²ºD (+) 21.0 - 23.5 Degree
EP/BP-Ninhydrin-Positive Substances-Each ≤ 0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤ 0.5 %
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Ammonium (NH₄) ≤ 0.02 %
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying ≤ 0.5 %
EP/BP - Ash (sulfated) ≤ 0.1 %
JP - Assay (C₆H₁₄N₄O₂HCl) (dried basis) 98.5 - 101.0 %
JP - Identification Passes Test
JP - Optical Rotation (+) 21.5 - 23.5 Degree
JP - pH (1 in 10) 4.7 - 6.2
JP - Clarity and Color of Solution Passes Test
JP - Sulfate (SO₄) ≤ 0.028 %
JP - Ammonium (NH₄) ≤ 0.02 %
JP - Heavy Metals (as Pb) ≤ 20 ppm
JP - Arsenic (As) ≤ 2 ppm
JP - Related Substances Passes Test
JP - Loss on Drying at 105°C ≤ 0.20 %
JP - Residue after Ignition ≤ 0.10 %
Endotoxin Concentration, IU/mg, For Information Only
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR