Agarose, OmniPur®

2010-100GM 2010-100GM
EM-2010EA 630.07 USD
CA-EM2010 EM-2010
Agarose, OmniPur®
Agarose

For resolution of small DNA fragments of less than 1000bp and PCR products. Prevents smearing or high fluorescence backgrounds. DNase-, RNase-, and protease-free.

Melting Pt: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://us.vwr.com/stibo/web/std.lang.all/43/75/23914375.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR