Water with 0.1% (v/v) Acetic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™

NP467-20 467-4 NP467-20 467-4
BJNP467-20EA 4762.12 USD
BJNP467-20 CABJ467-4 BJ467-4 CABJNP467-20
Water with 0.1% (v/v) Acetic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Water with 0.1% (v/v) Acetic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.9982 g/cm3
Storage Temperature: Ambient
EINECS: 200-580-7

Order Now

Specification Test Results

Acetic Acid (v/v) 0.09 - 0.11%
UV Cutoff 193 nm
Residue 1 mg/L
Color (APHA) 10
HPLC Gradient Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR