Aluminium oxide 50% in water, colloidal dispersion with dispersant, NanoArc® X1121W

Synonyms: Aluminium(III) oxideAlumina C

44925-36 44925-36 44925-22 44925-22
AA44925-36EA 117.27 USD
AA44925-36 AA44925-22 CAAA44925-36 CAAA44925-22
Aluminium oxide 50% in water, colloidal dispersion with dispersant, NanoArc® X1121W
Aluminium oxide

45nm APS for dry powder. Water-borne dispersion suitable for incorporation in water-compatible coatings.


Applications: Aluminum oxide water borne dispersion suitable for incorporation in water compatible coatings

Formula: Al₂O₃
MW: 101.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR