VWR® 8-Well Beta Rack
Supplier: VWR
VWR®
HS28208V
82010-758EA 100.47 USD
82010-758
VWR® 8-Well Beta Rack
Boxes Beta/Gamma Radiation Boxes
Ideal for safe transportation or benchtop storage of beta-emitting radioactive isotope samples.
  • Sturdy and snug transportation
  • Secure fit lids
Made of 8mm (5/16") thick acrylic, this sturdy and solid rack holds eight 1.5–2.0 mL microcentrifuge tubes. Snug lid ensures secure fit.