1-Heptadecanol 97%

Supplier: Ambeed
Warning

Synonyms: Heptadecan-1-olHexadecyl carbinol

A329132-1G A329132-10G A329132-250MG A329132-25G A329132-100G
77239-728EA 311 USD
77239-728 77239-730 77239-734 77239-732 77239-736
1-Heptadecanol 97%
1-Heptadecanol

Caution: for Research Use Only

Formula: H₃C(CH₂)₁₅CH₂OH
MW: 256.47 g/mol
MDL Number: MFCD00002822
CAS Number: 1454-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR