4-Isopropoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Isopropoxybenzeneboronic acid

A275207-1G A275207-25G A275207-10G A275207-100G A275207-5G
77250-402EA 11 USD
77250-402 77250-400 77250-398 77250-396 77250-394
4-Isopropoxyphenylboronic acid 97%
4-Isopropoxyphenylboronic acid

Caution: for Research Use Only

Formula: C₉H₁₃BO₃
MW: 180.01 g/mol
MDL Number: MFCD03427051
CAS Number: 153624-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR