2-Mercapto-5-methoxybenzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxy-1H-benzo[d]imidazole-2-thiol5-Methoxybenzimidazole-2-thiol2-Mercapto-5-methoxy-1H-benzimidazole5-Methoxy-2-benzimidazolethiol1,3-Dihydro-5-methoxy-2H-benzimidazole-2-thione

A245891-100G A245891-500G A245891-25G A245891-1000G
77276-176EA 76 USD
77276-176 77276-172 77276-174 77276-170
2-Mercapto-5-methoxybenzimidazole 98%
2-Mercapto-5-methoxybenzimidazole

Caution: for Research Use Only

Formula: C₈H₈N₂OS
MW: 180.23 g/mol
MDL Number: MFCD00134581
CAS Number: 37052-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR