4-Bromobutyl acetate 98%

Supplier: Ambeed
Warning

Synonyms: Acetic acid 4-bromobutyl ester

A217982-25G A217982-5G A217982-100G
77288-134EA 92 USD
77288-134 77288-136 77288-138
4-Bromobutyl acetate 98%
4-Bromobutyl acetate

Caution: for Research Use Only

Formula: C₆H₁₁BrO₂
MW: 195.06 g/mol
MDL Number: MFCD00000263
CAS Number: 4753-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR