1-Butylpyridiniumhexafluorophosphate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Butylpyridin-1-ium hexafluorophosphate(V)

A188823-100G A188823-1G A188823-5G A188823-25G A188823-10G
77236-790EA 94 USD
77236-790 77236-792 77236-794 77236-796 77236-798
1-Butylpyridiniumhexafluorophosphate 98%
1-Butylpyridinium hexafluorophosphate

Caution: for Research Use Only

Formula: C₉H₁₄F₆NP
MW: 281.18 g/mol
MDL Number: MFCD03093297
CAS Number: 186088-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR