Daidzein 98%

Supplier: Ambeed
Warning

Synonyms: 4',7-DihydroxyisoflavoneDaidzeol7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one

A129224-500G A129224-25G A129224-10G A129224-1G
77234-430EA 13 USD
77234-430 77234-436 77234-432 77234-434
Daidzein 98%
Daidzein

Caution: for Research Use Only

Formula: C₁₅H₁₀O₄
MW: 254.24 g/mol
MDL Number: MFCD00016954
CAS Number: 486-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR