3-Mercapto-1-hexanol 98%

Supplier: Ambeed
Danger

Synonyms: 3-Mercaptohexan-1-ol

A116052-5G
77271-196EA 33 USD
77271-196
3-Mercapto-1-hexanol 98%
3-Mercapto-1-hexanol

Caution: for Research Use Only

Formula: C₆H₁₄OS
MW: 134.24 g/mol
MDL Number: MFCD00792515
CAS Number: 51755-83-0
UN: 3334
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR