2-Aminothiobenzamide 97%

Supplier: Ambeed
Warning

Synonyms: 2-Aminobenzothioamide

A809630-5G A809630-250MG A809630-25G A809630-1G
77199-692EA 22 USD
77199-692 77199-696 77199-694 77199-698
2-Aminothiobenzamide 97%
2-Aminothiobenzamide

Caution: for Research Use Only

Formula: C₇H₈N₂S
MW: 152.22 g/mol
MDL Number: MFCD00963496
CAS Number: 2454-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR