3-Methoxythiobenzamide 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methoxybenzenecarbothioamide3-Methoxybenzothioamide

A747445-1G A747445-5G A747445-25G
77171-030EA 74.16 USD
77171-030 77171-026 77171-028
3-Methoxythiobenzamide 98%
3-Methoxythiobenzamide

Caution: for Research Use Only

Formula: C₈H₉NOS
MW: 167.23 g/mol
MDL Number: MFCD04627361
CAS Number: 64559-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR