1,10-Decanedithiol 98.5+%

Supplier: Ambeed
Danger

Synonyms: 1,10-Dimercaptodecane

A376318-5G
77195-548EA 16 USD
77195-548
1,10-Decanedithiol 98.5+%
1,10-Decanedithiol

Caution: for Research Use Only

Formula: C₁₀H₂₂S₂
MW: 206.42 g/mol
MDL Number: MFCD00022095
CAS Number: 1191-67-9
UN: 3335
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR