2-Acetylfuran 98%

Supplier: Ambeed
Danger

Synonyms: 2-Furyl methyl ketone1-(Furan-2-yl)ethanone

A221807-100G A221807-500G A221807-10G A221807-1000G
77152-704EA 88 USD
77152-704 77152-706 77152-710 77152-708
2-Acetylfuran 98%
2-Acetylfuran

Caution: for Research Use Only

Formula: C₆H₆O₂
MW: 110.11 g/mol
MDL Number: MFCD00003242
CAS Number: 1192-62-7
UN: 2811
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR