2-Fluoro-5-iodobenzaldehyde 98%

Supplier: Ambeed
Danger

A200354-25G
77139-316EA 88.58 USD
77139-316
2-Fluoro-5-iodobenzaldehyde 98%
2-Fluoro-5-iodobenzaldehyde

Caution: for Research Use Only

Formula: C₇H₄FIO
MW: 250.01 g/mol
MDL Number: MFCD03093999
CAS Number: 146137-76-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR