2-Chloro-5-iodotoluene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-4-iodo-2-methylbenzene

A191631-5G A191631-10G A191631-1G
77139-046EA 5.15 USD
77139-046 77139-048 77139-044
2-Chloro-5-iodotoluene 97%
2-Chloro-5-iodotoluene

Caution: for Research Use Only

Formula: C₇H₆ClI
MW: 252.48 g/mol
MDL Number: MFCD03093903
CAS Number: 116632-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR