2-Aminobenzenesulfonamide 98%

Supplier: Ambeed
Warning

A554211-25G A554211-1G
77116-942EA 11.33 USD
77116-942 77116-944
2-Aminobenzenesulfonamide 98%
2-Aminobenzenesulphonamide

Caution: for Research Use Only

Formula: C₆H₈N₂O₂S
MW: 172.21 g/mol
MDL Number: MFCD00007932
CAS Number: 3306-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR