2-Chloro-5-(trifluoromethyl)phenol 98%

Supplier: Ambeed
Danger

Synonyms: 4-Chloro-3-hydroxybenzotrifluoride1-Chlor-2-hydroxy-4-(trifluormethyl)benzol

A356342-5G
77115-204EA 10.61 USD
77115-204
2-Chloro-5-(trifluoromethyl)phenol 98%
2-Chloro-5-(trifluoromethyl)phenol

Caution: for Research Use Only

Formula: C₇H₄ClF₃O
MW: 196.56 g/mol
MDL Number: MFCD00019995
CAS Number: 40889-91-6
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR