2-Amino-4-ethylpyridine 96%

Supplier: Ambeed
Warning

Synonyms: 4-Ethyl-2-pyridylamine4-Ethylpyridin-2-amine

A197631-5G A197631-10G A197631-100G A197631-25G A197631-250MG A197631-1G
77112-036EA 70.04 USD
77112-036 77112-038 77112-040 77112-042 77112-044 77112-034
2-Amino-4-ethylpyridine 96%
2-Amino-4-ethylpyridine

Caution: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00129025
CAS Number: 33252-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR