3-Amino-6-bromo-4-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Amino-2-bromo-4-methylpyridine
A105075-100G
A105075-1G
A105075-250MG
A105075-5G
A105075-25G
A105075-10G
77107-500EA
289.63
USD
77107-500
77107-502
77107-504
77107-506
77565-834
77684-953
3-Amino-6-bromo-4-methylpyridine 98%
3-Amino-6-bromo-4-methylpyridine
Caution: For research use only.
Formula:
C₆H₇BrN₂ MW: 187.04 g/mol |
MDL Number:
MFCD08277280 CAS Number: 156118-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...