4-Ethylbenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-Ethylbenzene-1-sulfonyl chloride4-Ethylbenzene-1-sulphonyl chloride

A754699-5G A754699-1G A754699-25G
77006-272EA 101.97 USD
77006-272 77006-270 77006-274
4-Ethylbenzenesulfonyl chloride 98%
4-Ethylbenzenesulphonyl chloride

Caution: for Research Use Only

Formula: C₈H₉ClO₂S
MW: 204.68 g/mol
MDL Number: MFCD00041480
CAS Number: 16712-69-9
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR