4'-Hydroxyheptanophenone 98%

Supplier: Ambeed
Warning

A144771-1000G A144771-10G
76997-016EA 6.18 USD
76997-016 76997-014
4'-Hydroxyheptanophenone 98%
4'-Hydroxyheptanophenone

Caution: for Research Use Only

Formula: C₁₃H₁₈O₂
MW: 206.2800 g/mol
MDL Number: MFCD00191523
CAS Number: 14392-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR