2-Bromo-5-nitro-6-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-methyl-5-nitropyridine6-Bromo-2-methyl-3-nitropyridine

A143446-250MG A143446-5G A143446-1G A143446-100G A143446-10G A143446-25G
76996-960EA 5.15 USD
76996-960 76996-962 76996-964 76996-954 76996-956 76996-958
2-Bromo-5-nitro-6-picoline 98%
2-Bromo-5-nitro-6-picoline

Caution: for Research Use Only

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
MDL Number: MFCD03095219
CAS Number: 22282-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR