2-Chloronicotinoyl chloride 95%

Supplier: Ambeed
Danger

A391504-5G A391504-25G
76978-948EA 17.51 USD
76978-948 76978-946
2-Chloronicotinoyl chloride 95%
2-Chloronicotinoyl chloride

Caution: for Research Use Only

Formula: C₆H₃Cl₂NO
MW: 176.00 g/mol
MDL Number: MFCD00051677
CAS Number: 49609-84-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR