5-Bromo-3-nitro-o-toluic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-methyl-3-nitrobenzoic acid

55272-25G
76933-708EA 103 USD
76933-708
5-Bromo-3-nitro-o-toluic acid ≥97% research grade
5-Bromo-3-nitro-o-toluic acid

Caution: for Research Use Only

Formula: C₈H₆BrNO₄
MW: 260.0400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07357316
CAS Number: 107650-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR