2-Fluoro-5-iodobenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

23194-25G
76899-010EA 103 USD
76899-010
2-Fluoro-5-iodobenzaldehyde ≥97% research grade
2-Fluoro-5-iodobenzaldehyde

Caution: for Research Use Only

Formula: C₇H₄FIO
MW: 250.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093999
CAS Number: 146137-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR