3-Bromo-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-trifluoromethylbenzaldehyde

15865-25G
76908-328EA 60.77 USD
76908-328
3-Bromo-5-(trifluoromethyl)benzaldehyde ≥97%
3-Bromo-5-(trifluoromethyl)benzaldehyde

Caution: for Research Use Only

Formula: C₈H₄BrF₃O
MW: 253.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059506
CAS Number: 477535-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR