4-(Methylsulfonyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-(Methanesulphonyl)phenylboronic acid4-(Methanesulfonyl)phenylboronic acid4-(Methylsulphonyl)benzeneboronic acid

10276-25G
76906-334EA 60.77 USD
76906-334
4-(Methylsulfonyl)phenylboronic acid ≥97% research grade
4-(Methylsulphonyl)phenylboronic acid

Caution: for Research Use Only

Formula: C₇H₉BO₄S
MW: 200.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01630820
CAS Number: 149104-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR