4-Bromo-3-chlorobenzonitrile ≥95%

Supplier: AOBChem USA

Warning

21654-25G
76927-488EA 92.7 USD
76927-488
4-Bromo-3-chlorobenzonitrile ≥95%
4-Bromo-3-chlorobenzonitrile

Caution: for Research Use Only

Formula: C₇H₃BrClN
MW: 216.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834777
CAS Number: 57418-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR