5-Bromo-2-chloro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

13210-25G
76924-826EA 164.8 USD
76924-826
5-Bromo-2-chloro-4-iodopyridine ≥97%
5-Bromo-2-chloro-4-iodopyridine

Caution: for Research Use Only

Formula: C₅H₂BrClIN
MW: 318.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972189
CAS Number: 401892-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR